Order Entry
Export
ContactUsLinkComponent
 
Anti-TRP1 Mouse Monoclonal Antibody [clone: SPM456]
  ANTIA253433-100
New Product
Anti-TRP1 Mouse Monoclonal Antibody [clone: SPM456]
  ANTIA253433-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Glycoprotein 75
  • Antigen symbol:
    TRP1
  • Clonality:
    Monoclonal
  • Clone:
    SPM456
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17643
  • Antigen synonyms:
    CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|GP75|CATB|b-PROTEIN|5,6 dihydroxyindole 2 carboxylic acid oxidase|DHICA oxidase|5
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    75 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    SK-MEL-23 cells
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [SPM456] antibody to TRP1 for Flow Cytometry and IF with samples derived from human and mouse.

  • Validated Applications: Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: SPM456
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human;Mouse