Anti-p53 Mouse Recombinant Antibody [clone: rTP53/1739]
Catalog # ANTIA253355-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen NY-CO-13
- Antigen symbol:p53
- Clonality:Recombinant
- Clone:rTP53/1739
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P04637
- Antigen synonyms:P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:53 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human TP53 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rTP53/1739] antibody to p53 for WB and IHC-P with samples derived from human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: p53
Clonality: Recombinant
Clone: rTP53/1739
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human