Order Entry
Export
ContactUsLinkComponent
Anti-CD90 Mouse Monoclonal Antibody [clone: F15-42-1]
Anti-CD90 Mouse Monoclonal Antibody [clone: F15-42-1]
Catalog # ANTIA253307-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD90 Mouse Monoclonal Antibody [clone: F15-42-1]
Catalog # ANTIA253307-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD90
  • Clonality:
    Monoclonal
  • Clone:
    F15-42-1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Monkey,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04216
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    18 - 35 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Purified human brain CD90 protein
  • Tested applications:
    Functional studies
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [F15-42-1] antibody to CD90 for Flow Cytometry, IF and Functional Studies with samples derived from human andmonkey.

  • Validated Applications: Flow Cytometry, IF, Functional Studies
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 2 to 4 µg/ml

Type: Primary
Antigen: CD90
Clonality: Monoclonal
Clone: F15-42-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Monkey