Anti-PAX7 Rabbit Recombinant Antibody [Clone: PAX7/7079R]
ANTIA278628-100
New Product
- Antibody type:Primary
- Antigen name:FLJ37460
- Antigen symbol:PAX7
- Clonality:Recombinant
- Clone:PAX7/7079R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P23759
- Antigen synonyms:Paired box gene 7|HUP1|Paired box 7|OTTHUMP00000002534|PAX7|Paired box protein Pax-7|Paired domain gene HuP1|Paired box homeotic gene 7|Paired domain gene 7
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:57 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 301-505, of human PAX7 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [PAX7/7079R] antibody to PAX7 for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: PAX7
Clonality: Recombinant
Clone: PAX7/7079R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human