Order Entry
Export
ContactUsLinkComponent
Anti-MSH6 Rabbit Recombinant Antibody [Clone: MSH6/7065R]
Anti-MSH6 Rabbit Recombinant Antibody [Clone: MSH6/7065R]
  ANTIA278581-100
New Product
undefined
Anti-MSH6 Rabbit Recombinant Antibody [Clone: MSH6/7065R]
  ANTIA278581-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    DNA mismatch repair protein Msh6
  • Antigen symbol:
    MSH6
  • Clonality:
    Recombinant
  • Clone:
    MSH6/7065R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P52701
  • Antigen synonyms:
    HSAP|G/T mismatch-binding protein|hMSH6|HNPCC5|GTBP|G/T mismatch binding protein|HNPCC 5|MSH 6|GTMBP
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    163 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 374-540, of human MSH6 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [MSH6/7065R] antibody to MSH6 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: MSH6
Clonality: Recombinant
Clone: MSH6/7065R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human