Anti-ICAM1 Mouse Monoclonal Antibody [Clone: ICAM1/6917]
ANTIA278239-100
New Product
- Antibody type:Primary
- Antigen name:Antigen identified by monoclonal
- Antigen symbol:ICAM1
- Clonality:Monoclonal
- Clone:ICAM1/6917
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P05362
- Antigen synonyms:Cell surface glycoprotein P3.58 |Human rhinovirus receptor |CD 54 |BB2|CD54 antigen |CD54|CD_antigen=CD54|BB 2|ICAM 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:85-115 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment corresponding to human ICAM1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [ICAM1/6917] antibody to ICAM1 for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: ICAM1/6917
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human