Anti-HOXB2 Mouse Monoclonal Antibody [Clone: PCRP-HOXB2-1F2]
ANTIA278232-100
New Product
- Antibody type:Primary
- Antigen name:homeo box 2H
- Antigen symbol:HOXB2
- Clonality:Monoclonal
- Clone:PCRP-HOXB2-1F2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P14652
- Antigen synonyms:Homeobox protein Hox-2H|Hox 2 8|HOX2H|Homeobox protein Hox-2.8|Homeobox protein Hox-B2|HOX2|homeo box B2|homeobox B2|Hox 2H
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:37,9 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 114-220, of human HOXB2 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PCRP-HOXB2-1F2] antibody to HOXB2 for ELISA, IP, Flow Cytometry, IF, WB and IHC-P with samples derived from human.
- Validated Applications: ELISA, IP, Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HOXB2
Clonality: Monoclonal
Clone: PCRP-HOXB2-1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human