Anti-p53 Rabbit Recombinant Antibody [Clone: TP53/7002R]
Catalog # ANTIA278073-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen NY-CO-13
- Antigen symbol:p53
- Clonality:Recombinant
- Clone:TP53/7002R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P04637
- Antigen synonyms:P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:53 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human TP53 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [TP53/7002R] antibody to p53 for Flow Cytometry and IHC-P with samples derived from human.
- Validated Applications: Flow Cytometry, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: p53
Clonality: Recombinant
Clone: TP53/7002R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human