Anti-Growth Hormone Mouse Recombinant Antibody [Clone: rGH/4887]
Catalog # ANTIA277888-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:gH
- Antigen symbol:Growth Hormone
- Clonality:Recombinant
- Clone:rGH/4887
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P01241
- Antigen synonyms:GH-N|Growth hormone, normal|Growth hormone 1|Growth Hormone|GHB5|Growth hormone B5|GHN|GH1|Growth hormone, pituitary
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:20 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human Growth Hormone.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rGH/4887] antibody to Growth Hormone for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Growth Hormone
Clonality: Recombinant
Clone: rGH/4887
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human