Anti-Biotin Mouse Monoclonal Antibody [Clone: BTN/36]
Catalog # ANTIA277869-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Biotin
- Antigen symbol:Biotin
- Clonality:Monoclonal
- Clone:BTN/36
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Biotin
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:244Da
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Biotinylated sheep immunoglobulin.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [BTN/36] antibody to Biotin for Flow Cytometry, IF, WB and IHC-P.
- Validated Applications: Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 0,5 to 1 µg/million cells, IF: 0,5 to 1 µg/ml, WB: 0,5 to 2 µg/ml, IHC-P: 0,5 to 1 µg/ml
Type: Primary
Antigen: Biotin
Clonality: Monoclonal
Clone: BTN/36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: