Anti-Cytokeratin 17 Mouse Monoclonal Antibody [Clone: KRT17/4604]
Catalog # ANTIA277688-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin-17
- Antigen symbol:Cytokeratin 17
- Clonality:Monoclonal
- Clone:KRT17/4604
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q04695
- Antigen synonyms:keratin 17 epitope S2|CK-17|Cytokeratin 17|Keratin|39.1|K1C17_HUMAN|keratin 17 epitope S1|K17|CK 17
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:46 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human Cytokeratin 17.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [KRT17/4604] antibody to Cytokeratin 17 for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 17
Clonality: Monoclonal
Clone: KRT17/4604
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human