Anti-p63 Mouse Monoclonal Antibody [Clone: PANp63-6.1]
Catalog # ANTIA254416-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:AIS
- Antigen symbol:p63
- Clonality:Monoclonal
- Clone:PANp63-6.1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9H3D4
- Antigen synonyms:DN p63 alpha 1|id:ibd3516|B(p51A)|EEC3|CUSP|Chronic ulcerative stomatitis protein|B(p51B)|Amplified in squamous cell carcinoma|DNp63
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human p63 protein.
- Tested applications:IHC-P, ICC
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PANp63-6.1] antibody to p63 for WB, IHC-P, ICC and IP with samples derived from mouse, rat and human.
- Validated Applications: WB, IHC-P, ICC, IP
- Recommended Dilutions: WB: 1 µg/ml
Type: Primary
Antigen: p63
Clonality: Monoclonal
Clone: PANp63-6.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Human