Anti-IPO-38 Mouse Monoclonal Antibody [Clone: SPM260]
Catalog # ANTIA254368-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:IPO-38
- Clonality:Monoclonal
- Clone:SPM260
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Spleen cells of a patient with hairy cell leukemia.
- Tested applications:IHC-P
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [SPM260] antibody to IPO-38 for Flow Cytometry, IF and IHC-P with samples derived from human, mouse and rat.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: IPO-38
Clonality: Monoclonal
Clone: SPM260
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Mouse, Rat