Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 5 + 6 + 18 Mouse Monoclonal Antibody [Clone: SPM267]
Anti-Cytokeratin 5 + 6 + 18 Mouse Monoclonal Antibody [Clone: SPM267]
Catalog # ANTIA254140-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Cytokeratin 5 + 6 + 18 Mouse Monoclonal Antibody [Clone: SPM267]
Catalog # ANTIA254140-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Cytokeratin 5 + 6 + 18
  • Clonality:
    Monoclonal
  • Clone:
    SPM267
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Bovine,
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    45-58 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Detergent-insoluble fraction of psoriatic human epidermis.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [SPM267] antibody to Cytokeratin 5 + 6 + 18 for Flow Cytometry, IF, WB and IHC-P with samples derived from human, bovine and rat.

  • Validated Applications: Flow Cytometry, IF, WB, IHC-P
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 5 + 6 + 18
Clonality: Monoclonal
Clone: SPM267
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Bovine, Rat