Anti-CD47 Mouse Monoclonal Antibody [Clone: IAP/964 + B6H12.2]
ANTIA253910-100
New Product
- Antibody type:Primary
- Antigen name:Antigen identified by monoclonal
- Antigen symbol:CD47
- Clonality:Monoclonal
- Clone:IAP/964 + B6H12.2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:Multiple
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q08722
- Antigen synonyms:IAP|CD47_HUMAN|CD47|CD 47|CD47 antigen|CD47 molecule|Antigenic surface determinant protein OA3|CD47 glycoprotein|CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:~50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Clone IAP/964: Recombinant human IAP protein. Clone B6H12.2: Intact CD47 purified from placenta.
- Tested applications:Functional studies
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [IAP/964 + B6H12.2] antibody to CD47 for Functional Studies, WB, Flow Cytometry and IF with samples derived from human.
- Validated Applications: Functional Studies, WB, Flow Cytometry, IF
- Recommended Dilutions: WB: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: IAP/964 + B6H12.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human