Anti-CD36 Mouse Monoclonal Antibody [Clone: 185-1G2]
ANTIA253866-100
New Product
- Antibody type:Primary
- Antigen name:Adipocyte membrane protein
- Antigen symbol:CD36
- Clonality:Monoclonal
- Clone:185-1G2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P16671
- Antigen synonyms:CHDS7|CD36|BDPLT10|CD36 molecule (thrombospondin receptor)|CD36 molecule|Cluster determinant 36|CD36 antigen|CD36_HUMAN|CD36 antigen (collagen type I receptor, thrombospondin receptor)
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:80-90 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Stimulated human leukocytes.
- Tested applications:Functional studies
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [185-1G2] antibody to CD36 for Functional Studies, Flow Cytometry and IF with samples derived from human.
- Validated Applications: Functional Studies, Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 185-1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human