Anti-CD34 Mouse Monoclonal Antibody [Clone: QBEnd/10 + HPCA1/763]
ANTIA253860-100
New Product
- Antibody type:Primary
- Antigen name:CD34
- Antigen symbol:CD34
- Clonality:Monoclonal
- Clone:QBEnd/10 + HPCA1/763
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:Multiple
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P28906
- Antigen synonyms:CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:90-110 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Clone QBEnd/10: Detergent solubilized vesicular suspension prepared from human term placenta. Clone HPCA1/763: Recombinant full-length human HPCA1 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [QBEnd/10 + HPCA1/763] antibody to CD34 for Flow Cytometry, IF and IHC-P with samples derived from human.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 0,25 to 0,5 µg/ml
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEnd/10 + HPCA1/763
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human