Order Entry
Export
ContactUsLinkComponent
Anti-MUC16 Rabbit Recombinant Antibody [Clone: OCA125/4505R]
Anti-MUC16 Rabbit Recombinant Antibody [Clone: OCA125/4505R]
Catalog # ANTIA253808-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-MUC16 Rabbit Recombinant Antibody [Clone: OCA125/4505R]
Catalog # ANTIA253808-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CA 125
  • Antigen symbol:
    MUC16
  • Clonality:
    Recombinant
  • Clone:
    OCA125/4505R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8WXI7
  • Antigen synonyms:
    MUC 16|MUC-16|CA-125|CA125 ovarian cancer antigen|MUC16_HUMAN|Cancer antigen 125|MUC16|FLJ14303|CA125
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    >2,000 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to amino acids within 12,201-12,300 of CA125 protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [OCA125/4505R] antibody to MUC16 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: MUC16
Clonality: Recombinant
Clone: OCA125/4505R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human