Anti-Metabotropic Glutamate Receptor 7 Rabbit Polyclonal Antibody
Catalog # ANTIA96390-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:FLJ40498
- Antigen symbol:Metabotropic Glutamate Receptor 7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q14831
- Antigen synonyms:MGLU7|mGluR7|Metabotropic Glutamate Receptor 7|GLUR7|Glutamate receptor metabotropic 7|GRM7|OTTHUMP00000206961|GRM7_HUMAN|GPRC1G
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:102 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human GRM7 (amino acids 351-400)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to GRM7 for WB, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:40000
Type: Primary
Antigen: Metabotropic Glutamate Receptor 7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat