Anti-MiTF Rabbit Polyclonal Antibody
Catalog # ANTIA96381-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:bHLHe32
- Antigen symbol:MiTF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O75030
- Antigen synonyms:Homolog of mouse microphthalmia|Microphthalmia, mouse, homolog of|Microphthalmia associated transcription factor|CMM8|Class E basic helix-loop-helix protein 32|MiTF|Microphthalmia-associated transcription factor|mitfa|Mi
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:58 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human MITF (amino acids 151-200)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to MITF for WB, IHC, IF and ELISA with samples derived from Human and Mouse.
- Validated applications: WB, IHC, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: MiTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse