Anti-ARHGEF9 Rabbit Polyclonal Antibody
Catalog # ANTIA96359-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:9630036L12Rik
- Antigen symbol:ARHGEF9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# O43307
- Antigen synonyms:EIEE8|ARHG9_HUMAN|Cdc42 guanine nucleotide exchange factor (GEF) 9|collybistin I|Collybistin|hPEM-2 collybistin|ARHGEF9|A230067K14|HPEM 2
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:60 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human ARHGEF9 (amino acids 399-448)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to ARHGEF9 for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: IHC, ELISA
- Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: ARHGEF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat