Anti-EGFR Rabbit Polyclonal Antibody
Catalog # ANTIA96209-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:9030024J15Rik
- Antigen symbol:EGFR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P00533
- Antigen synonyms:EGFR_HUMAN|EGF R|Cell growth inhibiting protein 40 |EGFR|Cell proliferation inducing protein 61 |Avian erythroblastic leukemia viral (v erb b) oncogene homolog |AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (verbb) oncogene homolog
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:134 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human EGFR (amino acids 1139-1188)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to EGFR for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:20000
Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat