Anti-beta Tubulin Mouse Monoclonal Antibody [clone: 1B12]
Supplier: Antibodies.com
New Product
Certificates
About this item
Mouse monoclonal (1B12) antibody to beta Tubulin for WB, ICC/IF and IHC with samples derived from Human, Monkey, Rat and Mouse.
- Validated applications: WB, ICC/IF, IHC
- Recommended dilutions: WB: 1:5000 to 1:10000, ICC/IF: 1:500
Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Monkey;Rat;Mouse
Specifications
- Catalog No:
- ANTIA85428-50
- ANTIA85428-100
- Antigen symbol:
- beta Tubulin
- beta Tubulin
- Antigen name:
- Beta 4 tubulin
- Beta 4 tubulin
- Antigen synonyms:
- MGC117247|beta Tubulin|M40|beta Ib tubulin|Class I beta tubulin|Beta1 tubulin|MGC16435|Beta 5 tubulin|OK/SW cl.56
- MGC117247|beta Tubulin|M40|beta Ib tubulin|Class I beta tubulin|Beta1 tubulin|MGC16435|Beta 5 tubulin|OK/SW cl.56
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1B12
- 1B12
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P02554
- UniprotID# P02554
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Pig brain tubulin preparation
- Pig brain tubulin preparation
- Form:
- Liquid
- Liquid
- Molecular weight:
- 50 kDa
- 50 kDa
- Purification:
- Immunogen affinity purification
- Immunogen affinity purification
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Supplied in phosphate buffered saline with 50% glycerol and 5mM sodium azide
- Supplied in phosphate buffered saline with 50% glycerol and 5mM sodium azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC