Anti-GAP43 Mouse Monoclonal Antibody [clone: 3H14]
Supplier: Antibodies.com
New Product
Certificates
About this item
Mouse monoclonal (3H14) antibody to GAP43 for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.
- Validated applications: WB, ICC/IF, IHC
- Recommended dilutions: WB: 1:5000, ICC/IF: 1:1000 to 1:5000, IHC: 1:1000 to 1:500
Type: Primary
Antigen: GAP43
Clonality: Monoclonal
Clone: 3H14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human;Rat;Mouse
Specifications
- Catalog No:
- ANTIA85393-100
- ANTIA85393-50
- Antigen symbol:
- GAP43
- GAP43
- Antigen name:
- Axonal membrane protein GAP 43
- Axonal membrane protein GAP 43
- Antigen synonyms:
- Axonal membrane protein GAP-43|Growth-associated protein 43|Calmodulin binding protein P 57|Nerve growth related peptide GAP43|Growth Associated Protein 43|Nerve Growth Related Peptide|GAP43|B 50|F1
- Axonal membrane protein GAP-43|Growth-associated protein 43|Calmodulin binding protein P 57|Nerve growth related peptide GAP43|Growth Associated Protein 43|Nerve Growth Related Peptide|GAP43|B 50|F1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 3H14
- 3H14
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P17677
- UniprotID# P17677
- Isotype:
- IgM
- IgM
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant full length human GAP43, expressed in and purified from E. coli
- Recombinant full length human GAP43, expressed in and purified from E. coli
- Form:
- Liquid
- Liquid
- Molecular weight:
- ~43 kDa (by SDS-PAGE)
- ~43 kDa (by SDS-PAGE)
- Purification:
- Immunogen affinity purification
- Immunogen affinity purification
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Supplied in phosphate buffered saline with 50% glycerol and 5mM sodium azide
- Supplied in phosphate buffered saline with 50% glycerol and 5mM sodium azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC