Anti-MEK1 + MEK2 Rabbit Polyclonal Antibody
Catalog # ANTIA96160-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:AA589381
- Antigen symbol:MEK1 + MEK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q02750
- Antigen synonyms:MAP kinase kinase 2|Dual specificity mitogen-activated protein kinase kinase 1|CFC3|ERK activator kinase 2|ERK activator kinase 1|EC 2.7.12.2|Dual specificity mitogen-activated protein kinase kinase 2|FLJ26075|MAP kinase kinase 1
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:43 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human MEK1 (amino acids 252-301)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to MEK1 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: MEK1 + MEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat