Order Entry
Export
ContactUsLinkComponent
 
Anti-Connexin 43/GJA1 Rabbit Polyclonal Antibody
Catalog # ANTIA95550-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Connexin 43/GJA1 Rabbit Polyclonal Antibody
Catalog # ANTIA95550-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Connexin 43
  • Antigen symbol:
    Connexin 43/GJA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17302
  • Antigen synonyms:
    Connexin 43/GJA1|Connexin-43|DFNB38|Gap junction 43 kDa heart protein|Cx43|Gap junction protein alpha 1 43kDa|Gap junction alpha-1 protein|Cx 43|CXA1_HUMAN
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    43 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human Connexin 43 (amino acids 332-381)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to Connexin 43 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000

Type: Primary
Antigen: Connexin 43/GJA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat