Order Entry
Export
ContactUsLinkComponent
Anti-pan-myc Rabbit Polyclonal Antibody
Anti-pan-myc Rabbit Polyclonal Antibody
Catalog # ANTIA94654-100
Supplier:  Antibodies.com
New Product
undefined
Anti-pan-myc Rabbit Polyclonal Antibody
Catalog # ANTIA94654-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Avian myelocytomatosis viral oncogene homolog
  • Antigen symbol:
    pan-myc
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01106
  • Antigen synonyms:
    Class E basic helix-loop-helix protein 39|Class E basic helix-loop-helix protein 38|Class E basic helix-loop-helix protein 37|bHLHe39|c Myc|c-Myc|bHLHe37|L Myc|bHLHe38
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    48 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human Myc around the phosphorylation site of Thr358 (amino acids 325-374)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to Myc (phospho Thr358) for IHC, IP and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: IHC, IP, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:20000

Type: Primary
Antigen: pan-myc
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat