Anti-EGFR Rabbit Polyclonal Antibody
Catalog # ANTIA94625-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:9030024J15Rik
- Antigen symbol:EGFR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P00533
- Antigen synonyms:EGFR_HUMAN|EGF R|EGFR|Avian erythroblastic leukemia viral (verbb) oncogene homolog|Cell growth inhibiting protein 40|AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (v erb b) oncogene homolog|Cell proliferation inducing protein 61
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:134 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human EGFR around the phosphorylation site of Thr678 (amino acids 651-700)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to EGFR (phospho Thr678) for WB, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:40000
Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat