Anti-pan-myc Rabbit Polyclonal Antibody
Catalog # ANTIA96442-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Avian myelocytomatosis viral oncogene homolog
- Antigen symbol:pan-myc
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P01106
- Antigen synonyms:Class E basic helix-loop-helix protein 39|Class E basic helix-loop-helix protein 38|Class E basic helix-loop-helix protein 37|bHLHe39|c Myc|c-Myc|bHLHe37|L Myc|bHLHe38
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:48 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human MYC (amino acids 340-389)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to MYC for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:20000
Type: Primary
Antigen: pan-myc
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat