Order Entry
Export
ContactUsLinkComponent
Anti-SAE1 Rabbit Polyclonal Antibody
Anti-SAE1 Rabbit Polyclonal Antibody
Catalog # ANTIA96419-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-SAE1 Rabbit Polyclonal Antibody
Catalog # ANTIA96419-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Activator of SUMO1
  • Antigen symbol:
    SAE1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9UBE0
  • Antigen synonyms:
    SUA1|SAE1_HUMAN|SUMO 1 activating enzyme E1 N subunit|AOS1|Sentrin/SUMO activating protein AOS1|SUMO-activating enzyme subunit 1|HSPC140|SAE1|SUMO 1 activating enzyme subunit 1
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    38 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human AOS1 (amino acids 221-270)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to AOS1 for WB, IF and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IF, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:20000

Type: Primary
Antigen: SAE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat