Anti-Polyethylene Glycol Mouse Monoclonal Antibody (KLH)
AATB57000
New Product
- Antibody type:Secondary
- Antigen name:Polyethylene glycol
- Antigen symbol:PEG
- Clonality:Monoclonal
- Conjugation:KLH (Keyhole Limpet Hemocyanin)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Storage buffer:PBS, pH 7,2
- Storage temperature:Store at −29 °C
- Concentration:1 mg/ml
- Shipping temperature:20
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µG
PEG Antibody, mAb, Mouse has high affinity for the PEG backbone.
- Affinity purified
The antibody binds to the variety of PEG such as PEG40K, PEG20K, PEG5K, PEG12, PEGylated drugs and PEG conjugates.
: Some reagents such as Tween-20, Triton X-100 and NP-40 interfere with the PEG antibody interaction due to structural similarity to PEG.
Type: Secondary
Antigen: Polyethylene glycol
Clonality: Monoclonal
Clone:
Conjugation: KLH
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse, Human