Anti-CSA Rabbit Polyclonal Antibody
ANTIA310023-100
New Product
- Antibody type:Primary
- Antigen name:CSA
- Antigen symbol:CSA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Oryza sativa
- Western blot:Yes
- Form:Liquid
- Gene ID:4325181
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% thiomersal
- Molecular weight:50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of Oryza sativa CSA.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to CSA for WB with samples derived from Oryza sativa.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: CSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Oryza sativa