Anti-IL-21 Rabbit Monoclonal Antibody [clone: ARC60383]
Catalog # ANTIA309923-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CVID11
- Antigen symbol:IL-21
- Clonality:Monoclonal
- Clone:ARC60383
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9HBE4
- Antigen synonyms:IL21|Interleukin 21|Interleukin-21|IL-21|Interleukin 21 isoform|IL21_HUMAN|IL 21|Cytokine CX2|DN 452A22.7
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
- Molecular weight:18 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of human IL21.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC60383] antibody to IL-21 for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 1:1,000 to 1:5,000
Type: Primary
Antigen: IL-21
Clonality: Monoclonal
Clone: ARC60383
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human