Anti-TB1 Rabbit Polyclonal Antibody
Catalog # ANTIA309900-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:HMSN6B
- Antigen symbol:TB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Oryza sativa
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q8LN68
- Antigen synonyms:S2546_HUMAN|TB1|S2546|SLC25A46|Solute carrier family 25 member 46
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% thiomersal
- Molecular weight:52 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of Oryza sativa OsTB1.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to TB1 for WB with samples derived from Oryza sativa.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: TB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Oryza sativa