Order Entry
Export
ContactUsLinkComponent
Anti-IRE1 Rabbit Polyclonal Antibody
Anti-IRE1 Rabbit Polyclonal Antibody
Catalog # ANTIA309829-100
Supplier:  Antibodies.com
New Product
undefined
Anti-IRE1 Rabbit Polyclonal Antibody
Catalog # ANTIA309829-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Endoplasmic reticulum (ER) to nucleus signalling 1
  • Antigen symbol:
    IRE1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Oryza sativa
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q7XIT1
  • Antigen synonyms:
    ERN1_HUMAN|Endoribonuclease|Endoplasmic reticulum-to-nucleus signaling 1|hIRE1p|ERN 1|Endoplasmic reticulum to nucleus signaling 1|hIRE 1p|ER to nucleus signaling 1|Ern1
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,02% sodium azide
  • Molecular weight:
    90 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant protein of Oryza sativa OsIRE1.
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to IRE1 for WB with samples derived from Oryza sativa.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2,000

Type: Primary
Antigen: IRE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Oryza sativa