Anti-Connexin 43/GJA1 Rabbit Monoclonal Antibody [clone: ARC58819]
Catalog # ANTIA309695-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Connexin 43
- Antigen symbol:Connexin 43 / GJA1
- Clonality:Monoclonal
- Clone:ARC58819
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P17302
- Antigen synonyms:Connexin-43|DFNB38|Connexin 43 / GJA1|Gap junction 43 kDa heart protein|Cx43|Gap junction protein alpha 1 43kDa|Gap junction alpha-1 protein|Cx 43|CXA1_HUMAN
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
- Molecular weight:43 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthesised peptide derived from human Connexin 43.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC58819] antibody to Connexin 43/GJA1 for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:1,000 to 1:5,000
Type: Primary
Antigen: Connexin 43 / GJA1
Clonality: Monoclonal
Clone: ARC58819
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat