Anti-CD3 epsilon + CD3G Rabbit Monoclonal Antibody [clone: ARC5083-01]
Catalog # ANTIA309691-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD3 epsilon + CD3G
- Antigen symbol:CD3 epsilon + CD3G
- Clonality:Monoclonal
- Clone:ARC5083-01
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P07766, P09693
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
- Molecular weight:23 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-207(CD3E)& 1-375(CD3G) of human CD3 epsilon&CD3 gamma Heterodimer.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC5083-01] antibody to CD3 epsilon + CD3G for WB and flow cytometry with samples derived from human.
- Validated applications: WB, flow cytometry
- Recommended dilutions: WB: 1:1,000 to 1:5,000, flow cytometry: 1:50 to 1:200
Type: Primary
Antigen: CD3 epsilon + CD3G
Clonality: Monoclonal
Clone: ARC5083-01
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human