Anti-Proteasome 20S LMP7 Rabbit Polyclonal Antibody
Catalog # ANTIA309447-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:ALDD
- Antigen symbol:Proteasome 20S LMP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P28062
- Antigen synonyms:D6S216E|Low molecular mass protein 7|Macropain subunit C13|LMP 7|Large multifunctional protease 7|LMP7|Low molecular weight protein 7|Large multifunctional peptidase 7|D6S216
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Molecular weight:24 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of human Proteasome 20S LMP7.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Proteasome 20S LMP7 for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: Proteasome 20S LMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat