Anti-Nesprin3 Rabbit Monoclonal Antibody [clone: ARC3013]
ANTIA309420-100
New Product
- Antibody type:Primary
- Antigen name:C14orf49
- Antigen symbol:Nesprin3
- Clonality:Monoclonal
- Clone:ARC3013
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q6ZMZ3
- Antigen synonyms:Nesprin-3|NET53|Nesp3|MGC75267|Nuclear Envelope Spectrin Repeat Protein 3|Nesprin 3|Chromosome 14 open reading frame 49|FLJ25605|Nesprin3
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:112 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of Human Nesprin3.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit monoclonal [ARC3013] antibody to Nesprin3 for WB with samples derived from human and mouse.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: Nesprin3
Clonality: Monoclonal
Clone: ARC3013
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse