Order Entry
Export
ContactUsLinkComponent
 
Anti-Nanog Rabbit Monoclonal Antibody [clone: ARC58438]
  ANTIA309414-100
New Product
undefined
Anti-Nanog Rabbit Monoclonal Antibody [clone: ARC58438]
  ANTIA309414-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Embryonic stem cell specific homeobox protein (Nanog)
  • Antigen symbol:
    Nanog
  • Clonality:
    Monoclonal
  • Clone:
    ARC58438
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9H9S0
  • Antigen synonyms:
    ENK|NANOG_HUMAN|hNanog|Nanog homeobox|homeobox transcription factor Nanog-delta 48|Homeobox transcription factor Nanog|FLJ12581|Homeobox protein NANOG
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant protein of human Nanog.
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [ARC58438] antibody to Nanog for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:2,000 to 1:10,000

Type: Primary
Antigen: Nanog
Clonality: Monoclonal
Clone: ARC58438
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat