Anti-YKL-40/CHI3L1 Rabbit Polyclonal Antibody
ANTIA309216-100
New Product
- Antibody type:Primary
- Antigen name:YKL-40 / CHI3L1
- Antigen symbol:YKL-40 / CHI3L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P36222
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Molecular weight:43 kDa
- Sequence:YKLVCYYTSWSQYREGDGSCFPDALDRFLCTHIIYSFANISNDHIDTWEWNDVTLYGMLNTLKNRNPNLKTLLSVGGWNFGSQRFSKIASNTQSRRTFIKSVPPFLRTHGFDGLDLAWLYPGRRDKQHFTTLIKEMKAEFIKEAQPGKKQLLLSAALSAGKVTIDSSYDIAKISQHLDFISIMTYDFHGAWRGTTGHHSPLFRGQEDASPDRFSNTDYAVGYMLRLGAPASKLVMGIPTFGRSFTLASSETGVGAPISGPGIPGRFTKEAGTLAYYEICDFLRGATVHRILGQQVPYATKGNQWVGYDDQESVKSKVQYLKDRQLAGAMVWALDLDDFQGSFCGQDLRFPLTNAIKDALAAT
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 22-383 of human YKL-40/CHI3L1 (NP_001267.2).
- Tested applications:ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to YKL-40/CHI3L1 for WB and ICC/IF with samples derived from human and mouse.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:100 to 1:500, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: YKL-40 / CHI3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse