Anti-p53 (phospho Thr55) Rabbit Monoclonal Antibody [clone: ARC1530]
Catalog # ANTIA309163-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen NY-CO-13
- Antigen symbol:p53
- Clonality:Monoclonal
- Clone:ARC1530
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P04637
- Antigen synonyms:P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
- Modification:phospho Thr55
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:53 kDa
- Sequence:WFTED
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic phosphorylated peptide around T55 of human p53 (P04637).
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC1530] antibody to p53 (phospho Thr55) for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: ARC1530
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human