Order Entry
Export
ContactUsLinkComponent
 
Anti-CD44 Rabbit Polyclonal Antibody
  ANTIA309094-100
New Product
undefined
Anti-CD44 Rabbit Polyclonal Antibody
  ANTIA309094-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    BA-1
  • Antigen symbol:
    CD44
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P16070
  • Antigen synonyms:
    CD44_HUMAN|CD44|CDw44|CD44 molecule (Indian blood group)|chondroitin sulfate proteoglycan 8|CD 44|Cell surface glycoprotein CD44|CD44 antigen|CD44 molecule
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,02% sodium azide
  • Sequence:
    VNRDGTRYVQKGEYRTNPEDIYPSNPTDDDVSSGSSSERSSTSGGYIFYTFSTVHPIPDEDSPWITDSTDRIPATRDQDTFHPSGGSHTTHGSESDGHSH
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 148-247 of human CD44 (NP_001189486.1).
  • Tested applications:
    ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to CD44 for ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: ICC/IF
  • Recommended dilutions: ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: CD44
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat