Order Entry
Export
ContactUsLinkComponent
Anti-LSP1 Rabbit Monoclonal Antibody [clone: ARC1952]
Anti-LSP1 Rabbit Monoclonal Antibody [clone: ARC1952]
Catalog # ANTIA308852-100
Supplier:  Antibodies.com
New Product
undefined
Anti-LSP1 Rabbit Monoclonal Antibody [clone: ARC1952]
Catalog # ANTIA308852-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    47 kDa actin binding protein
  • Antigen symbol:
    LSP1
  • Clonality:
    Monoclonal
  • Clone:
    ARC1952
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P33241
  • Antigen synonyms:
    Leufactin|LSP 1|52 kDa phosphoprotein|Leukocyte F actin binding protein|47 kDa actin-binding protein|LSP1|F actin binding and cytoskeleton associated protein|Leukocyte specific protein 1|Leufactin (leukocyte F-actin binding protein)
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    47 kDa
  • Sequence:
    TAGRTPKLARQASIELPSMAVASTKSRWETGEVQAQSAAKTPSCKDIVAGDMSKKSLWEQKGGSKTSSTIKSTPSGKRYKFVATGHGKYEKVLVEGGPAP
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 240-339 of human LSP1 (P33241).
  • Tested applications:
    ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC1952] antibody to LSP1 for WB and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:2,000, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: LSP1
Clonality: Monoclonal
Clone: ARC1952
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat