Order Entry
Export
ContactUsLinkComponent
 
Anti-CD38 Rabbit Recombinant Antibody [clone: CD38/8335R]
  ANTIA314695-100
New Product
Anti-CD38 Rabbit Recombinant Antibody [clone: CD38/8335R]
  ANTIA314695-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Acute lymphoblastic leukemia cells antigen CD38
  • Antigen symbol:
    CD38
  • Clonality:
    Recombinant
  • Clone:
    CD38/8335R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28907
  • Antigen synonyms:
    ADPRC 1|ADP-ribosyl cyclase 1|CD38 antigen|ADPRC1|cADPr hydrolase 1|ADP ribosyl cyclase/cyclic ADP-ribose hydrolase|ADP ribosyl cyclase|ADP ribosyl cyclase 1|CD 38
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    ~45 kDa (Glycoprotein)/35 kDa (protein core)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 200-300) of human CD38. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [CD38/8335R] antibody to CD38 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: CD38
Clonality: Recombinant
Clone: CD38/8335R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human