Anti-Carboxypeptidase A Mouse Recombinant Antibody [clone: rCPA1/8518]
Catalog # ANTIA314557-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Carboxypeptidase A
- Antigen symbol:Carboxypeptidase A
- Clonality:Recombinant
- Clone:rCPA1/8518
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P15085
- Antigen synonyms:CBPA1_HUMAN|Procarboxypeptidase A1 pancreatic|carboxypeptidase A1 (pancreatic)|CPA|CPA1|Pancreatic carboxypeptidase A1|Carboxypeptidase A1|Carboxypeptidase A1 precursor
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:47 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CPA1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rCPA1/8518] antibody to Carboxypeptidase A for WB and IHC-P with samples derived from human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Carboxypeptidase A
Clonality: Recombinant
Clone: rCPA1/8518
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human