Order Entry
Export
ContactUsLinkComponent
Anti-MEK1 + MEK2 Rabbit Polyclonal Antibody
Anti-MEK1 + MEK2 Rabbit Polyclonal Antibody
  ANTIA308735-100
New Product
undefined
Anti-MEK1 + MEK2 Rabbit Polyclonal Antibody
  ANTIA308735-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    AA589381
  • Antigen symbol:
    MEK1 + MEK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q02750, P36507
  • Antigen synonyms:
    MAP kinase kinase 2|Dual specificity mitogen-activated protein kinase kinase 1|CFC3|ERK activator kinase 2|ERK activator kinase 1|EC 2.7.12.2|Dual specificity mitogen-activated protein kinase kinase 2|FLJ26075|MAP kinase kinase 1
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,02% sodium azide
  • Molecular weight:
    44 kDa
  • Sequence:
    SMANS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic phosphorylated peptide around S217 and S221 of human MAP2K2MAP2K1 (NP_002746.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to MEK1 + MEK2 for WB with samples derived from mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:100 to 1:500

Type: Primary
Antigen: MEK1 + MEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat