Order Entry
Export
ContactUsLinkComponent
Anti-MEK1 (phospho Thr292) Rabbit Polyclonal Antibody
Anti-MEK1 (phospho Thr292) Rabbit Polyclonal Antibody
Catalog # ANTIA308236-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-MEK1 (phospho Thr292) Rabbit Polyclonal Antibody
Catalog # ANTIA308236-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dual specificity mitogen activated protein kinase kinase 1
  • Antigen symbol:
    MEK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q02750
  • Antigen synonyms:
    MAPKK 1|Dual specificity mitogen-activated protein kinase kinase 1|MAP2K1|MAPKK1|MAP kinase/Erk kinase 1|MEK 1|ERK activator kinase 1|MAP kinase kinase 1|MAPK/ERK kinase 1
  • Modification:
    phospho Thr292
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% thiomersal
  • Molecular weight:
    45 kDa
  • Sequence:
    PRTPG
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic phosphorylated peptide around T292 of human MEK1 (NP_002746.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to MEK1 (phospho Thr292) for WB with samples derived from human and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat