Anti-MEK1 (phospho Thr292) Rabbit Monoclonal Antibody [clone: ARC1566]
ANTIA308235-100
New Product
- Antibody type:Primary
- Antigen name:Dual specificity mitogen activated protein kinase kinase 1
- Antigen symbol:MEK1
- Clonality:Monoclonal
- Clone:ARC1566
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q02750
- Antigen synonyms:MAPKK 1|Dual specificity mitogen-activated protein kinase kinase 1|MAP2K1|MAPKK1|MAP kinase/Erk kinase 1|MEK 1|ERK activator kinase 1|MAP kinase kinase 1|MAPK/ERK kinase 1
- Modification:phospho Thr292
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:45 kDa
- Sequence:PRTPG
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic phosphorylated peptide around T292 of human Phospho-MEK1-T292 (Q02750)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit monoclonal [ARC1566] antibody to MEK1 (phospho Thr292) for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: MEK1
Clonality: Monoclonal
Clone: ARC1566
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat