Order Entry
Export
ContactUsLinkComponent
 
Anti-ACE2 Rabbit Recombinant Antibody [clone: DM47]
New Product
 
undefined
Anti-ACE2 Rabbit Recombinant Antibody [clone: DM47]
New Product

 

  • Catalog No:
  • ANTIA318648-500
  • ANTIA318648-100
  • Antigen symbol:
  • ACE2
  • ACE2
  • Antigen name:
  • ACE 2
  • ACE 2
  • Antigen synonyms:
  • ACEH|Angiotensin converting enzyme homolog|Angiotensin Converting Enzyme 2|ACE2|Angiotensin I Converting Enzyme (peptidyl dipeptidase A) 2|ACE related carboxypeptidase|ACE-related carboxypeptidase|ACE2_HUMAN|Angiotensin converting enzyme like protein
  • ACEH|Angiotensin converting enzyme homolog|Angiotensin Converting Enzyme 2|ACE2|Angiotensin I Converting Enzyme (peptidyl dipeptidase A) 2|ACE related carboxypeptidase|ACE-related carboxypeptidase|ACE2_HUMAN|Angiotensin converting enzyme like protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Recombinant
  • Recombinant
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • DM47
  • DM47
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • UniprotID# Q9BYF1
  • UniprotID# Q9BYF1
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Lyophilised
  • Lyophilised
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Concentration:
  • Reconstitution dependent
  • Reconstitution dependent
  • Storage buffer:
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation
  • Storage temperature:
  • Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C